Eco stopcock plugs, without bore and taper 1:10
Supplier: Bohlender
J1200-18 J1200-29 J1200-14 J1200-12 J1200-10 J1200-24
BOHLJ1200-18EA 8 EUR
BOHLJ1200-18 BOHLJ1200-29 BOHLJ1200-14 BOHLJ1200-12 BOHLJ1200-10 BOHLJ1200-24
Eco stopcock plugs, without bore and taper 1:10
Stopcocks
Made of PFTE, white, with grip and nut of PP, red.
  • Universal chemical resistance
  • Temperature resistance: from −10 to +120 °C
  • Taper: 1:10
  • Without bore
  • Washer included