Niobium standard solution, 1000 mg/L Nb in water with hydrofluoric acid (max. 1%) standard for AAS
Supplier: AccuStandard
AA38W1 AA38W5
ACCUAA38W1EA 0 EUR
ACCUAA38W1 ACCUAA38W5
Niobium standard solution, 1000 mg/L Nb in water with hydrofluoric acid (max. 1%) standard for AAS
Standards Cationic Standards Niobium Standards