Ruthenium standard solution, 10,000 mg/L Ru in 10% hydrochloric acid standard for ICP
Supplier: AccuStandard
ICP48H10X05 ICP48H10X1 ICP48H10X5
ACCUICP48H10X05EA 0 EUR
ACCUICP48H10X05 ACCUICP48H10X1 ACCUICP48H10X5
Ruthenium standard solution, 10,000 mg/L Ru in 10% hydrochloric acid standard for ICP
Standards Cationic Standards Ruthenium Standards