Rubidium standard solution, 10,000 mg/L Rb in dil. nitric acid standard for ICP
Supplier: AccuStandard
ICP47N10X05 ICP47N10X1 ICP47N10X5
ACCUICP47N10X05EA 0 EUR
ACCUICP47N10X05 ACCUICP47N10X1 ACCUICP47N10X5
Rubidium standard solution, 10,000 mg/L Rb in dil. nitric acid standard for ICP
Standards Cationic Standards Rubidium Standards