Silicon standard solution, 1000 mg/L Si in water with hydrofluoric acid (max. 1%) (from (NH₄)₂SiF₆), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
38717.AD
38717.AE
38717.AP
38717.ADEA
83.8
EUR
38717.AD
38717.AE
38717.AP
Silicon standard solution, 1000 mg/L Si in water with hydrofluoric acid (max. 1%) (from (NH₄)₂SiF₆), standard for ICP, Specpure®
Standards
Cationic Standards
Silicium Standards
Molecular Formula: (NH₄)₂SiF₆ in H₂O/tr. F-
Physical Form: Liquid
MDL No.: MFCD00085311
Certifications: Traceable to NIST