Terbium standard solution, 10000 mg/L Tb in dil. nitric acid standard for ICP
Supplier: AccuStandard
ICP59N10X05 ICP59N10X1 ICP59N10X5
ACCUICP59N10X05EA 0 EUR
ACCUICP59N10X05 ACCUICP59N10X1 ACCUICP59N10X5
Terbium standard solution, 10000 mg/L Tb in dil. nitric acid standard for ICP
Standards Cationic Standards Terbium Standards