Boron trichloride 1 M in heptane
Supplier: Thermo Fisher Scientific
H32785.AE H32785.AP
H32785.AEEA 120 EUR
H32785.AE H32785.AP
Boron trichloride 1 M in heptane
Boron trichloride
CAS No.: 10294-33-4 Molecular Formula: BBr3 Formula Weight: 250.52 Flash Point: -4°(25°F) Boiling Point: 90° UN No.: 3384 MDL No.: MFCD00011312
Formula: BCl₃
MW: 117.17 g/mol
Boiling Pt: 90 °C
Melting Pt: -107
Density: 1.876
Flash Pt: –4 °C (25 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011313
CAS Number: 10294-34-5
EINECS: 233-658-4
UN: 3384
ADR: 6.1 I
Merck Index: 12,01378