Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Supplier: Thermo Fisher Scientific
Warning
Synonyms: DE, Diatomaceous earth, flux-calcined
H33152.0B H33152.0I
H33152.0BEA 63 EUR
H33152.0B H33152.0I
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Celite® 545, filter aid
CAS No.: 68855-54-9 MDL No.: MFCD00132803
Formula: SiO₂
MW: 60.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00132803
CAS Number: 68855-54-9
EINECS: 272-489-0