4-Bromothiobenzamide ≥97%

Supplier: Thermo Fisher Scientific
Danger

H55956.03 H55956.06
H55956.03EA 51.9 EUR
H55956.03 H55956.06
4-Bromothiobenzamide ≥97%
4-Bromothiobenzamide
Formula: C₇H₆BrNS
Melting Pt: 139…143 °C
Storage Temperature: Ambient
MDL Number: MFCD04114360
CAS Number: 26197-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR