Ammonium acetate 5 M in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Synonyms: Acetic acid ammonium salt
M-Clarity™ quality level = MQ200
09691-100ML 09691-1L 09691-250ML
SIAL09691-100MLEA 67.2 EUR
SIAL09691-100ML SIAL09691-1L SIAL09691-250ML
Ammonium acetate 5 M in aqueous solution, Sigma-Aldrich®
Ammonium acetate
Formula: H₃CCOONH₄
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM