4-Hexyloxyaniline ≥99%
Supplier: Thermo Fisher Scientific
Danger
H55868.06 H55868.14
H55868.06EA 75 EUR
H55868.06 H55868.14
4-Hexyloxyaniline ≥99%
4-Hexyloxyaniline
Formula: C₁₂H₁₉NO
Boiling Pt: 155…158 °C (5 mmHg)
Melting Pt: 43…45 °C
Flash Pt: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007868
CAS Number: 39905-57-2
EINECS: 254-696-8