Oxalyl dichloride 2 M in dichloromethane
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Oxalyl chloride
H31617.AE H31617.AP
H31617.AEEA 55.8 EUR
H31617.AE H31617.AP
Oxalyl dichloride 2 M in dichloromethane
Oxalyl dichloride
CAS No.: 79-37-8 Molecular Formula: C2Cl2O2 Formula Weight: 126.93 UN No.: 2922 MDL No.: MFCD00000704
Formula: C₂Cl₂O₂
MW: 126.93 g/mol
Boiling Pt: 63
Melting Pt: -12
Density: 1.335
Storage Temperature: Ambient
MDL Number: MFCD00000704
CAS Number: 79-37-8
EINECS: 201-200-2
UN: 2922
ADR: 8 II
Merck Index: 12,07046