2,5-Dibromohydroquinone ≥97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2,5-Dibromobenzene-1,4-diol
H55453.06 H55453.14
H55453.06EA 81.2 EUR
H55453.06 H55453.14
2,5-Dibromohydroquinone ≥97%
2,5-Dibromohydroquinone
Purity: 97% CAS No.: 14753-51-6 Molecular Formula: C6H4Br2O2 Formula Weight: 267.90 Melting Point: 191-194° MDL No.: MFCD00192664
Formula: C₆H₄Br₂O₂
MW: 267.9 g/mol
Melting Pt: 191…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00192664
CAS Number: 14753-51-6