2-Chloro-5-fluoropyrimidine ≥97%
Supplier: Thermo Fisher Scientific
Danger
H55055.03 H55055.06
H55055.03EA 76.3 EUR
H55055.03 H55055.06
2-Chloro-5-fluoropyrimidine ≥97%
2-Chloro-5-fluoropyrimidine
Formula: C₄H₂ClFN₂
Density: 1.073
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD03788197
CAS Number: 62802-42-0
UN: 3267
ADR: 8,III