Valeramide 97%

Supplier: Thermo Fisher Scientific
Warning

422440025 422440250 422440050
ACRO422440025EA 33.4 EUR
ACRO422440025 ACRO422440250 ACRO422440050
Valeramide 97%
Valeramide
Formula: C₅H₁₀O₂
Melting Pt: 101…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00041895
CAS Number: 626-97-1
EINECS: 210-974-0

Order Now

Specification Test Results

Appearance White Glistening powder or flakes
Infrared spectrum Conforms
Melting point 101 to 106 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR