Phthaloyl dichloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Phthaloyl chloride, o-Phthaloyl dichloride

M-Clarity™ quality level = MQ100

P40409-100G P40409-500G
SIALP40409-100GEA 52.2 EUR
SIALP40409-100G SIALP40409-500G
Phthaloyl dichloride, Sigma-Aldrich®
Phthaloyl dichloride
Formula: C₈H₄Cl₂O₂
MDL Number: MFCD00000666
CAS Number: 88-95-9
EINECS: 201-869-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR