Triethylborane 1 mol/l in hexane, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

195030-100ML 195030-800ML
SIAL195030-100MLEA 105 EUR
SIAL195030-100ML SIAL195030-800ML
Triethylborane 1 mol/l in hexane, Sigma-Aldrich®
Triethylborane
Formula: C₆H₁₅B
MW: 98 g/mol
MDL Number: MFCD00009022
CAS Number: 97-94-9
EINECS: 202-620-9
UN: 3399
ADR: 4.3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR