Silver p-toluenesulphonate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

176427-10G 176427-25G
SIAL176427-10GEA 148 EUR
SIAL176427-10G SIAL176427-25G
Silver p-toluenesulphonate, Sigma-Aldrich®
Silver p-toluenesulphonate
Formula: C₇H₇AgO₃S
MDL Number: MFCD00007511
CAS Number: 16836-95-6
EINECS: 240-859-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR