Sodium hippurate hydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
H9380-25G
H9380-100G
H9380-1KG
SIALH9380-25GEA
99.5
EUR
SIALH9380-25G
SIALH9380-100G
SIALH9380-1KG
Sodium hippurate hydrate, Sigma-Aldrich®
Sodium hippurate hydrate
Formula:
C₉H₈NNaO₃·H₂O |
CAS Number:
532-94-5 EINECS: 208-548-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...