(R)-(+)-1,1'-Bi-2-naphthol, Sigma-Aldrich®
Supplier: Merck
Synonyms: (R)-[1,1'-Binaphthalene]-2,2'-diol, (R)-BINOL, (R)-(+)-1,1'-Binaphthyl-2,2'-Diol
M-Clarity™ quality level = MQ100
246948-10G 246948-1G 246948-5G
SIAL246948-10GEA 229 EUR
SIAL246948-10G SIAL246948-1G SIAL246948-5G
(R)-(+)-1,1'-Bi-2-naphthol, Sigma-Aldrich®
(R)-(+)-1,1'-Bi-2-naphthol
Formula: C₂₀H₁₄O₂
MW: 286.33 g/mol
MDL Number: MFCD00004068
CAS Number: 18531-94-7
UN: 2811
ADR: 6.1 III