VWR®, Chemical Resistant Coats, Polyester/Hydro-Tec, Reusable
Supplier: VWR Collection
113-1829EA 122 EUR
113-1829 113-1827 113-1826 113-1825 113-1830 113-1828 113-1844 113-1838 113-1831 113-1832 113-1833 113-1834 113-1835 113-1837 113-1839 113-1840 113-1841 113-1842 113-1843 113-1836 113-5994 113-6115 113-6114 113-6110 113-6102 113-6103 113-6104 113-6100 113-6105 113-6106 113-6107 113-6116 113-6109 113-6101 113-6111 113-6112 113-6113 113-6099 113-6098 113-6097 113-6096 113-6108 113-6157 113-6147 113-6138 113-6139 113-6140 113-6141 113-6142 113-6143 113-6144 113-6146 113-6158 113-6148 113-6149 113-6150 113-6151 113-6152 113-6153 113-6154 113-6155 113-6156 113-6145 114-9322
VWR®, Chemical Resistant Coats, Polyester/Hydro-Tec, Reusable
Workwear
These anti-static coats are manufactured from 99% polyester fibres with 1% Hydro-Tec finish (carbon treated), which makes them both extremely durable and excellent for repelling dirt, oil, chemicals and water. All coats feature a stand-up collar, two side pockets and one breast pocket, all three protected by a flap.
  • Extremely durable thanks to plain weave construction
  • Innovative Hydro-Tec finish provides protection against splashes
  • Front closure with press studs covered by Velcro® flap
  • Women's style comes with a fitted waist
  • Washable at 60 °C
Colour: White, grey or blue
Density: 275 g/m²
EN 13034:2005 (type 6), EN 1149-5; complies with Oeko-Tex® Standard 100
Certifications: EN 13034:2005 (type 6), EN 1149-5; complies with Oeko-Tex® Standard 100