Sucrose solutions for refractometers
Supplier: Atago
RE-110030
RE-110050
RE-110010
RE-110060
RE-110020
RE-110040
CRE-110010
CRE-110050
ATAGRE-110030EA
0
EUR
ATAGRE-110030
ATAGRE-110050
ATAGRE-110010
ATAGRE-110060
ATAGRE-110020
ATAGRE-110040
ATAGCRE-110010
ATAGCRE-110050
Sucrose solutions for refractometers
Standards
Refractive Index Standards
Sucrose solutions are used for confirming the Brix measurements of refractometers.
- These solutions have a shelf life of 6 weeks