7-Phenylenanthic acid ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 97%
CAS No.: 40228-90-8
Molecular Formula: C6H5(CH2)6COOH
Formula Weight: 206.29
Flash Point:
Formula:
C₁₃H₁₈O₂ Boiling Pt: 147…150 °C (0.4 mmHg) Melting Pt: ∼ 20 °C Density: 1.009 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00037801 CAS Number: 40228-90-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...