Ethynyltributyltin 96%
Supplier: Thermo Fisher Scientific
|
Danger
|
L20178.03
L20178.06
L20178.03EA
70.2
EUR
L20178.03
L20178.06
Ethynyltributyltin 96%
Ethynyltributyltin
Purity: 96%
CAS No.: 994-89-8
Molecular Formula: C14H28Sn
Formula Weight: 315.09
Storage Temperature: 2°C to 8°C
Flash Point: 73°(163°F)
Refractive Index: 1.4770
Boiling Point: 70°/0.2mm
Beilstein Reference: 3537659
UN No.: 2788
MDL No.: MFCD00009420
Formula:
C₁₄H₂₈Sn Boiling Pt: 70 °C (0.2 mmHg) Density: 1.080 Flash Pt: 73 °C (163 °F) |
MDL Number:
MFCD00009420 CAS Number: 994-89-8 UN: 2788 ADR: 6.1,III |