Ethylmagnesium bromide 0.9 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
210471000 210478000
210471000.EA 58.3 EUR
210471000. 210478000.
Ethylmagnesium bromide 0.9 M in THF, AcroSeal®
Ethylmagnesium bromide
Formula: C₂H₅BrMg
MW: 133.27 g/mol
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00000043
CAS Number: 925-90-6
EINECS: 213-127-3
UN: 3399
ADR: 4.3 I
Order Now
Specification Test Results
Appearance Brown to grey-brown to brown-black Solution (May develop some turbidity or precipitate)
Titration Grignard 0.85 to 1.15 M