2-Imidazolidinone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
I601-1KG
I601-25G
I601-500G
SIALI601-1KGEA
521
EUR
SIALI601-1KG
SIALI601-25G
SIALI601-500G
2-Imidazolidinone, Sigma-Aldrich®
2-Imidazolidinone
Formula:
C₃H₆N₂O |
MDL Number:
MFCD00005257 CAS Number: 120-93-4 EINECS: 204-436-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...