Dimethylamine 2 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
391956-100ML 391956-800ML
SIAL391956-100MLEA 96.5 EUR
SIAL391956-100ML SIAL391956-800ML
Dimethylamine 2 mol/l in THF, Sigma-Aldrich®
Dimethylamine
Formula: C₂H₇N
MW: 45.08 g/mol
Boiling Pt: 6.9
Melting Pt: -92.8
Density: 0.938
Flash Pt: -55
Storage Temperature: Ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 2924
ADR: 3 II
Merck Index: 12,03278