Potassium dicyanoargentate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

379166-10G 379166-50G
SIAL379166-10GEA 105 EUR
SIAL379166-10G SIAL379166-50G
Potassium dicyanoargentate, Sigma-Aldrich®
Potassium dicyanoargentate
Formula: C₂AgKN₂
MDL Number: MFCD00036297
CAS Number: 506-61-6
EINECS: 208-047-0
UN: 1588
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR