Phenylmagnesium bromide 2.9 mol/l, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
703575-100ML 703575-800ML
SIAL703575-100MLEA 127 EUR
SIAL703575-100ML SIAL703575-800ML
Phenylmagnesium bromide 2.9 mol/l, Sigma-Aldrich®
Phenylmagnesium bromide
Formula: C₆H₅BrMg
MW: 181.31 g/mol
MDL Number: MFCD00000038
CAS Number: 100-58-3
EINECS: 202-867-2
UN: 2924
ADR: 3 II