Terpinolene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

86485-100ML 86485-500ML
SIAL86485-100MLEA 45.4 EUR
SIAL86485-100ML SIAL86485-500ML
Terpinolene, Sigma-Aldrich®
Terpinolene
Formula: C₁₀H₁₆
MDL Number: MFCD00049191
CAS Number: 586-62-9
EINECS: 209-578-0
UN: 2541
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR