2-(Tributylstannyl)pyridine tech. 80%, Technical Grade
Supplier: Thermo Fisher Scientific
|
Danger
|
B24911.03
B24911.06
B24911.14
B24911.03EA
51.5
EUR
B24911.03
B24911.06
B24911.14
2-(Tributylstannyl)pyridine tech. 80%, Technical Grade
2-(Tributylstannyl)pyridine
Purity: 80%
CAS No.: 17997-47-6
Molecular Formula: C17H31NSn
Formula Weight: 368.15
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5130
Boiling Point: 130-132°/0.8mm
Beilstein Reference: 479329
UN No.: 2788
MDL No.: MFCD00052052
Formula:
C₁₇H₃₁NSn Boiling Pt: 130…132 °C (0.8 mmHg) Density: 1.15 Storage Temperature: Refrigerator |
MDL Number:
MFCD00052052 CAS Number: 17997-47-6 UN: 2788 ADR: 6.1,III |