2-Methylbenzyl mercaptan
Supplier: Thermo Scientific
Synonyms:
2-Methylbenzylmercaptan, o-Xylene-α-thiol
TL00468.1
TL00468.10
TL00468.50
MAYBTL00468.1EA
0
EUR
MAYBTL00468.1
MAYBTL00468.10
MAYBTL00468.50
2-Methylbenzyl mercaptan
2-Methylbenzyl mercaptan
Formula:
C₈H₁₀S Boiling Pt: 52 °C (1 mmHg) Melting Pt: 208…210 °C Density: 1.034 g/cm³ (20 °C) |
MDL Number:
MFCD00022074 CAS Number: 7341-24-4 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...