Ampicillin sodium salt, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

M-Clarity™ quality level = MQ200

A9518-25G A9518-5G A9518-100G
SIALA9518-25GEA 255 EUR
SIALA9518-25G SIALA9518-5G SIALA9518-100G
Ampicillin sodium salt, Sigma-Aldrich®
Ampicillin sodium salt
Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3
EINECS: 200-708-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR