Ampicillin sodium salt, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt
M-Clarity™ quality level = MQ200
A0166-100G A0166-25G A0166-5G
SIALA0166-100GEA 2290 EUR
SIALA0166-100G SIALA0166-25G SIALA0166-5G
Ampicillin sodium salt, Sigma-Aldrich®
Ampicillin sodium salt
Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3
EINECS: 200-708-1