Tetracycline hydrochloride
Supplier: Roth Carl
Warning
0237.1 0237.2 0237.3
ROTH0237.1EA 81.3 EUR
ROTH0237.1 ROTH0237.2 ROTH0237.3
Tetracycline hydrochloride
Tetracycline hydrochloride
Formula: C₂₂H₂₅ClN₂O₈
Storage Temperature: Ambient
MDL Number: MFCD00078142
CAS Number: 64-75-5
EINECS: 200-593-8