Tetracycline hydrochloride, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
T8032-10MG T8032-20MG
SIALT8032-10MGEA 49 EUR
SIALT8032-10MG SIALT8032-20MG
Tetracycline hydrochloride, Sigma-Aldrich®
Tetracycline hydrochloride
Formula: C₂₂H₂₅ClN₂O₈
Storage Temperature: Ambient
MDL Number: MFCD00078142
CAS Number: 64-75-5
EINECS: 200-593-8