Knitted gloves, Nitras® 03400, partially coated, with knitted cuff
Supplier: AS ARBEITSSCHUTZ
034008 034009 0340010 0340011 034007
112-2298EA 1.9 EUR
112-2298 112-2299 112-2364 112-2369 ASAS034007
Knitted gloves, Nitras® 03400, partially coated, with knitted cuff
Gloves
100% cotton jersey knit gloves with 3/4 nitrile coating.
  • High wearing comfort and flexibility
  • With knitted waistband
Total length: 240mm
Ideal for construction, industry and commerce.
Certifications: EN 388 (4111X); Cat. II