Knitted gloves, Nitras® 03430, partially coated, with cuff
Supplier: AS ARBEITSSCHUTZ
034309 0343010 0343111
112-2405EA 2.8 EUR
112-2405 112-2406 112-2407
Knitted gloves, Nitras® 03430, partially coated, with cuff
Gloves
Work gloves made of 100% cotton jersey, partially coated with nitrile rubber (NBR).
  • Offers great comfort and flexibility
  • With safety cuff
  • Good manual dexterity enables effective work
Ideal for the automotive industry, manufacturing companies or construction trades.
EN 388 4221
Certifications: EN 388 (4221X); Cat. II