Knitted gloves, Nitras® 03440, fully coated, with cuff
Supplier: AS ARBEITSSCHUTZ
034409 0344010 0344111
112-2408EA 2.9 EUR
112-2408 112-2409 112-2414
Knitted gloves, Nitras® 03440, fully coated, with cuff
Gloves
Heavy work gloves made of 100% cotton jersey, fully coated with nitrile rubber.
  • Offers great comfort and flexibility
  • With safety cuff
  • Fully coated for maximum protection
For use in the automotive industry, in manufacturing plants or in the construction trade.
EN 388 4221
Certifications: EN 388 (4221X); Cat. II