Knitted gloves, Nitras® 6100, with PVC knobs
Supplier: AS ARBEITSSCHUTZ
610010 61009
112-2622EA 18.3 EUR
112-2622 112-2621
Knitted gloves, Nitras® 6100, with PVC knobs
Gloves
Economical 10 gauge nylon/cotton knit gloves.
  • Blue PVC nubs on palm for better grip
  • With knitted cuff for the perfect finish on the wrist
For use in general work that requires a good grip.
Total length: 250mm
Certifications: EN 388 (2142X); Cat. II