Cut resistant gloves, Nitras® TAEKI5® 6730
Supplier: AS ARBEITSSCHUTZ
6730S 6730M 6730L 6730XL 67302XL 67303XL
ASAS6730SEA 67 EUR
ASAS6730S ASAS6730M 112-2635 112-2636 112-2637 112-2638
Cut resistant gloves, Nitras® TAEKI5® 6730
Gloves
Knitted gloves made of TAEKI5® special cut protection fiber knit, without coating.
  • Extremely high cut resistance
  • Good abrasion resistance, heat resistant
  • Seamless processing
  • Also suitable as under gloves
Ideal for cutting work in the metal and automotive industries where good touch is also required.
Farbe: grau
EN 388 4540
Certifications: EN 388 (3X42C), EN 407 (X1XXXX); Cat. II
Approved for food contact