Cotton Linen Glove
Supplier: TRENKA
100T7 100T8 100T9
112-2647EA 6.3 EUR
112-2647 112-2648 112-2649
Cotton Linen Glove
Gloves
Interlock gloves made of 75% polyamide and 25% cotton. Ideal for delicate, dry work; when assembling small parts or when handling.
  • Proven quality, particularly hard-wearing
  • Doubleface binding
  • fingers with layers; comfortable to wear
Gesamtlänge: 250 mm
EN 388 xx4x
Certifications: EN 388 (XX4X)