General purpose gloves, Maxifoam
Supplier: TRENKA
130321 130314 130291 130307
112-2651EA 6.1 EUR
112-2651 112-2650 112-2653 112-2654
General purpose gloves, Maxifoam
Gloves
Flexible, lightweight and seamless knitted nylon glove (13 gauge) with knit wrist. Partially dipped with nitrile foam.
  • Very comfortable to wear
  • Extremely sure-grip, abrasion-resistant
Very good when working with oily and greasy materials.
EN 388 4131
Certifications: EN 388 (4131)