Supplier: RENTEX
130758 130760 130748 130750 130752 130754 130756
RENT130758EA 0 EUR
RENT130758 RENT130760 RENT130748 RENT130750 RENT130752 RENT130754 RENT130756
Workwear
Chemical resistant, flame retardant dungarees made from 94% Nomex® III, 5% Para-Aramid and 1% metal. The material is also permanently antistatic. Increased wearing comfort due to light fabric.
  • Elastic straps, large bib pocket with snap button, waistband adjustable with jeans button
  • Two slanted pockets on the front, two jeans pockets with flaps on the back, ruler pocket
  • Decorative seams in combination colour
  • Fly with metal zip, covered on the inside
Weight: 260 g/m²
Certifications: EN 531 A, B1, C1; EN 533; EN 470-1; EN 1149-1