Jacket with waistband, Trenkatex® by filia, Proban® SF, BD 3311
Supplier: RENTEX
85548 85550 85552 85554 85556 85558
RENT85548EA 0 EUR
RENT85548 RENT85550 RENT85552 RENT85554 RENT85556 RENT85558
Jacket with waistband, Trenkatex® by filia, Proban® SF, BD 3311
Workwear
Proban® welding protection jacket made from Tecapro, 100% cotton, 3/1 twill. Protects against convection and radiated heat.
  • Two breast pockets with press stud, one of which is a mobile phone pocket
  • One breast pocket inside, two concealed side pockets, two large inner pockets
  • Concealed press stud panel, decorative seams in combination colour
  • Adjustable cuffs
Weight: 320 g/m²
Certifications: EN 531 A, B1, C1; EN 533; EN 470-1