Supplier: KIND ARBEITSSICHERHEIT
82430M 82430L 824301XL 824302XL 82426M 82426L 824261XL 824262XL
113-08368EA 0 EUR
113-08368 113-08367 113-08365 113-08366 113-08364 113-08363 113-08361 113-08362
Workwear
High visibility jacket with rain and flame retardant properties, made from 98% polyester and 2% carbon fibres. The coating is 100% polyurethane, hydrophilic and breathable. The COTTEX® lining has the highest flame retardant index 3.
  • Flame retardant, anti-static, chemical proof, waterproof and breathable
  • Removable flame retardant body and sleeve thermal lining with inner pocket
  • Hood (adjustable) with drawstring and peak in the collar
  • Two-way front zip to collar end, Velcro cover
  • Inside cuffs, knitted waistband as a windbreak, Velcro straps for width adjustment
  • Two chest and side pockets, one zip pocket
  • reflective trim
Length approx. 90 cm
Certifications: EN 343 3-2,  EN 471 2-2, EN 1149-5 (1149-3), EN 13034, EN ISO 14116