High Visibility Trousers, Rhine
Supplier: ORBIT
GB3FWTXS GB3FWTS GB3FWTM GB3FWTL GB3FWT1XL GB3FWT2XL GB3FWT3XL GB3FWT4XL GB3FWTRXS GB3FWTRS GB3FWTRM GB3FWTRL GB3FWTR1XL GB3FWTR2XL GB3FWTR3XL GB3FWTR4XL
113-08135EA 205 EUR
113-08135 113-08134 113-08125 113-08124 113-08120 113-08121 113-08122 113-08123 113-08133 113-08132 113-08131 113-08130 113-08126 113-08127 113-08128 113-08129
High Visibility Trousers, Rhine
Workwear
Round trousers with elastic band.
  • 3-layer Gore-Tex® Benbecula fabric, windproof, waterproof and breathable
  • Lower leg narrowing with zipper
  • Oversize surcharge: size 4XL +25%
Certifications: EN 471 (1-2); EN 343 (3-3)