Functional T-shirts, Match Coolmax®
Supplier: FRISTADS KANSAS
1004715402XL 100471540L 100471540M 100471540S 100471540XL 100471540XS 1004719002XL 100471900L 100471900M 100471900S 100471900XL 100471900XS 1004719302XL 100471930L 100471930M 100471930S 100471930XL 100471930XS 1004719402XL 100471940L 100471940M 100471940S 100471940XL 100471940XS
KWIN1004715402XLEA 37.9 EUR
KWIN1004715402XL KWIN100471540L KWIN100471540M KWIN100471540S KWIN100471540XL KWIN100471540XS KWIN1004719002XL KWIN100471900L KWIN100471900M KWIN100471900S KWIN100471900XL KWIN100471900XS KWIN1004719302XL KWIN100471930L KWIN100471930M KWIN100471930S KWIN100471930XL KWIN100471930XS KWIN1004719402XL KWIN100471940L KWIN100471940M KWIN100471940S KWIN100471940XL KWIN100471940XS
Functional T-shirts, Match Coolmax®
Workwear
Functional shirt made of 50% Coolmax® Dacron fibre and 50% polyester. Transports sweat away from the body and dries quickly.
  • Moisture wicking
  • Crew neck
  • Side vents, lengthened back
Weight: 140 g/m²