Safety Boots, Wellingtons
Supplier: FELDTMANN HELMUT
3537037 3537638 3537639 3537640 3537641 3537642 3537643 3537644 3537645 3537646 3537647 3537648
114-3492EA 0 EUR
114-3492 114-3493 114-3494 114-3495 114-3496 114-3497 114-3498 114-3499 114-3500 114-3501 114-3502 114-3503
Safety Boots, Wellingtons
Shoes and Boots
PVC/Nitrile safety boots with cotton fabric lining. Resistant to animal fats, oils and acids, antistatic.
  • Very flexible even at low temperatures
  • Steel toe cap and corrosion-free steel midsole
Colour: Black
width: 11
Shaft height: 380 mm