Safety shoes, lace-up, SN 10
Supplier: ATLAS - the shoe company
41139 41140 41141
114-1367EA 0 EUR
114-1367 114-1368 114-1369
Safety shoes, lace-up, SN 10
Shoes and Boots
Lace-up shoe made from SOFT nubuck leather and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Skin friendly, abrasion resistant aktiv-X lining for enhanced breathability
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Protective steel toecap (200 J)
Colour: Brown
Certifications: DGUV 112-191; EN ISO 20345; S2, SRC